Aldehyde resin on polystyrene (PS), beads 330-1225 µm, 15-50 mesh Loading: 1.0-1.2 mmol/g
Supplier: Supra Sciences
Certificates
About this item
:
Specification Test Results
- Colour:Off-white
- Form:Granule
- Loading:1.0-1.2 mmol/g
- Particle size:330-1225 μm (15-50 mesh)