HRV 3C Protease Activity Assay Kit (Colorimetric), BioVision Inc.
75833-276
: Avantor
:
- Description:HRV 3C Protease Activity Assay Kit (Colorimetric)
- Size:100 assays
- Sample Type:Purified proteins and cell samples.
- Detection Method:Absorbance (OD= 405 nm)
- Shelf Life:12 Months
- Cat. No.:75833-276
- Storage temperature:–20 °C
- Supplier no.:K214-100