ContactUsLinkComponent
Biopsy Punch Tray, Sklar
Biopsy Punch Tray, Sklar
Catalog # 75818-008
Supplier:  Sklar
undefined
Biopsy Punch Tray, Sklar
Catalog # 75818-008
Supplier:  Sklar
Supplier Number:  96-1712
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Biopsy Punch Tray
  • Cat. No.:
    75818-008
  • Packaging:
    25/Case
Specifications
About this item
Biopsy Punch Trays are industry standard
Each tray includes the following instruments and prep materials: 4mm Biopsy Punch, Webster Needle Holder, Curved Iris Scissors, Tissue Forceps, 30ga x 1/2" Needle, 25ga x 5/8" Needle, 3cc Syringe, Alcohol Prep Pad, PVP Ointment, (4) Gauze Sponges (2x2", 12-ply), Fenestrated Drape (18x26"), (2) Band-Aids, Gloves, and Tray.