Order Entry
United States
ContactUsLinkComponent
Single Element Standards for ICP, Zirconium, Agilent Technologies
Single Element Standards for ICP, Zirconium, Agilent Technologies
Catalog # ULICP-040
Supplier:  Ultra Scientific
Small Business Enterprise
CAS Number:  
undefined
Single Element Standards for ICP, Zirconium, Agilent Technologies
Catalog # ULICP-040
Supplier:  Ultra Scientific
Supplier Number:  ICP-040
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    5% HNO₃
  • Size:
    125 ml
  • Cat. No.:
    ULICP-040
  • Element:
    Zirconium
  • Supplier no.:
    ICP-040

Specifications

About this item

The ICP standard is manufactured and verified under Ultra’s ISO 9001 registered quality system

It is traceable to NIST standard reference materials (SRM) whenever possible.

ULTRAgrade® documentation data includes elemental identity, trace metal impurities, and final element concentration by ICP; traceability data to NIST SRM by both wet chemical assay and ICP on final solution lot; and final solution density.

It is prepared from starting materials having a purity of 99.999% (where possible), high purity acids, and ASTM Type I water (18 megaohm).