Anti-HIF-1-alpha Rabbit Monoclonal Antibody [clone: RM242]
Catalog # 103255-316
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Primary
- Antigen Symbol:HIF-1-alpha
- Clonality:Monoclonal
- Clone:Clone: RM242
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Format:Purified
- Form:Liquid
- Gene ID:3091
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Shipping Temperature:4°C
- Immunogen:A peptide corresponding to Hypoxia-inducible factor 1-alpha
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-316
- Supplier no.:31-1121-00
Specifications
About this item
This antibody reacts to human HIF-1-alpha (Hypoxia-inducible factor 1-alpha).
Immunohistochemistry (IHC): 1:500-1:1000 dilution;
Western Blot (WB): 1:1000-1:2000 dilution.