Order Entry
United States
ContactUsLinkComponent
 
Anti-RSK1 Rabbit Monoclonal Antibody [clone: RM233]
  103255-606
undefined
Anti-RSK1 Rabbit Monoclonal Antibody [clone: RM233]
  103255-606
 :  31-1117-00

 

  • Antibody Type:
    Primary
  • Antigen Name:
    phospho-Rsk1
  • Antigen Symbol:
    RSK1
  • Clonality:
    Monoclonal
  • Clone:
    Clone: RM233
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Format:
    Purified
  • Form:
    Liquid
  • Gene ID:
    6195 , 6196
  • Modification:
    Thr359/Ser363
  • Storage Buffer:
    50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
  • Storage Temperature:
    -20.0°C
  • Shipping Temperature:
    4°C
  • Immunogen:
    A phospho-peptide corresponding to human phospho-Rsk1 (Thr359/Ser363)
  • Purification:
    Protein A affinity purified from an animal origin-free culture supernatant
  • Cat. No.:
    103255-606
  • Supplier no.:
    31-1117-00

 

 

This antibody reacts to human RSK1 only when phosphorylated at Thr359/Ser363.

Immunohistochemistry (IHC): 1:200-1:500 dilution;
Western Blot (WB): 1:1000-1:2000 dilution.