Anti-5-hmC Rabbit Monoclonal Antibody [clone: RM236]
Catalog # 103255-546
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Primary
- Antigen Name:5-hydroxymethylcytosine
- Antigen Symbol:5-hmC
- Clonality:Monoclonal
- Clone:Clone: RM236
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:50 µg
- ChIP:Yes
- Format:Purified
- Form:Liquid
- Antigen Synonyms:5'-hmC
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Concentration:1 mg/mL
- Shipping Temperature:4°C
- Immunogen:BSA-conjugated 5-hydroxymethylcytosine.
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-546
- Supplier no.:31-1111-00
Specifications
About this item
This antibody reacts to 5-hydroxymethylcytosine in both single-stranded and double-stranded DNA. No cross reactivity with non-methylated cytosine and methylcytosine in DNA.
Dot Blot: 0.2 ug/mL – 1 ug/mL;
ELISA: 0.1 ug/mL - 1 ug/mL;
IHC: 0.1 ug/mL - 1 ug/mL.