Anti-H2AX Rabbit Monoclonal Antibody [clone: RM224]
103255-526
- Antibody Type:Primary
- Antigen Name:Phospho-Gamma Histone H2AX (Ser139)
- Antigen Symbol:H2AX
- Clonality:Monoclonal
- Clone:Clone: RM224
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:100 µg
- Epitope:Phosphorylated Gamma Serine 139 in Histone H2AX
- Format:Purified
- Form:Liquid
- Gene ID:3014
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Concentration:1 mg/mL
- Shipping Temperature:4°C
- Immunogen:A phospho-peptide corresponding to Phospho-Histone H2AX (Ser139)
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-526
- Supplier no.:31-1104-00
This antibody reacts to Histone H2A.X only when phosphorylated at serine 139. No cross reactivity with other phosphorylated histones.
Western Blot: 0.5 ug/mL – 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 1 ug/mL;
ICC: 0.5 ug/mL - 2 ug/mL.