Order Entry
United States
ContactUsLinkComponent
Anti-H3T3p Rabbit Monoclonal Antibody [clone: RM159]
Anti-H3T3p Rabbit Monoclonal Antibody [clone: RM159]
Catalog # 103255-510
Supplier:  RevMAb Biosciences
undefined
Anti-H3T3p Rabbit Monoclonal Antibody [clone: RM159]
Catalog # 103255-510
Supplier:  RevMAb Biosciences
Supplier Number:  31-1072-00

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Phospho-Histone H3 (Thr3), H3T3p
  • Antigen Symbol:
    H3T3p
  • Clonality:
    Monoclonal
  • Clone:
    Clone: RM159
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Multiple
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    Phosphorylated Threonine 3 in Histone H3
  • Format:
    Purified
  • Form:
    Liquid
  • Gene ID:
    8350
  • Modification:
    Thr3
  • Storage Buffer:
    50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
  • Storage Temperature:
    -20.0°C
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4°C
  • Immunogen:
    A phospho-peptide corresponding to Phospho-Histone H3 (Thr3)
  • Purification:
    Protein A affinity purified from an animal origin-free culture supernatant
  • Cat. No.:
    103255-510
  • Supplier no.:
    31-1072-00

Specifications

About this item

This antibody reacts to Histone H3 phosphorylated at Threonine 3. No cross reactivity with other phosphorylated histones

Western Blot: 0.1 ug/mL - 1 ug/mL;
ICC: 0.5 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 1 ug/mL.