Order Entry
United States
ContactUsLinkComponent
Anti-H4K16ac Rabbit Monoclonal Antibody [clone: RM204]
Anti-H4K16ac Rabbit Monoclonal Antibody [clone: RM204]
Catalog # 103255-496
Supplier:  RevMAb Biosciences
undefined
Anti-H4K16ac Rabbit Monoclonal Antibody [clone: RM204]
Catalog # 103255-496
Supplier:  RevMAb Biosciences
Supplier Number:  31-1083-00

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Acetylated Histone H4 Lysine 16, H4K16ac
  • Antigen Symbol:
    H4K16ac
  • Clonality:
    Monoclonal
  • Clone:
    Clone: RM204
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Multiple
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    Acetyllysine 16 in Histone H4
  • Format:
    Purified
  • Form:
    Liquid
  • Gene ID:
    121504
  • Storage Buffer:
    50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
  • Storage Temperature:
    -20.0°C
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4°C
  • Immunogen:
    An acetyl-peptide corresponding to the Acetyl-Histone H4 (Lys16).
  • Purification:
    Protein A affinity purified from an animal origin-free culture supernatant
  • Cat. No.:
    103255-496
  • Supplier no.:
    31-1083-00

Specifications

About this item

This antibody reacts to Histone H4 acetylated at Lysine 16 (K16ac). No cross reactivity with other acetylated Lysines in Histone H4.

Western Blot: 0.5 ug/mL - 2 ug/mL;
ELISA: 0.5 ug/mL - 1 ug/mL;
Multiplex: 0.5 ug/mL – 2 ug/mL.