Anti-H3K9me2 Rabbit Monoclonal Antibody [clone: RM151]
Catalog # 103255-466
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Primary
- Antigen Name:Dimethylated Histone H3 Lysine 9, H3K9me2
- Antigen Symbol:H3K9me2
- Clonality:Monoclonal
- Clone:Clone: RM151
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:100 µg
- Epitope:Dimethyllysine 9 in Histone H3
- ChIP:Yes
- Format:Purified
- Form:Liquid
- Gene ID:8350
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Concentration:1 mg/mL
- Shipping Temperature:4°C
- Immunogen:A dimethyl-peptide corresponding to Dimethyl-Histone H3 (Lys9).
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-466
- Supplier no.:31-1059-00
Specifications
About this item
This antibody reacts to Histone H3 dimethylated at Lysine 9 (K9me2). No cross reactivity with monomethylated Lysine 9 (K9me1) or trimethylated Lysine 9 (K9me3), or other methylation in histone H3.
Western Blot: 0.25 ug/mL – 1 ug/mL;
ChIP: 2 ug/mL- 10 ug/mL;
ICC: 0.5 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.