Anti-H3K18ac Rabbit Monoclonal Antibody [clone: RM166]
Catalog # 103255-462
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Primary
- Antigen Name:Acetylated Histone H3 Lysine 18, H3K18ac
- Antigen Symbol:H3K18ac
- Clonality:Monoclonal
- Clone:Clone: RM166
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:100 µg
- Epitope:Acetyllysine 18 in Histone H3
- ChIP:Yes
- Format:Purified
- Form:Liquid
- Gene ID:8350
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Concentration:1 mg/mL
- Shipping Temperature:4°C
- Immunogen:An acetyl-peptide corresponding to the Acetyl-Histone H3 (Lys18)
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-462
- Supplier no.:31-1055-00
Specifications
About this item
This antibody reacts to Histone H3 acetylated at Lysine 18 (K18ac). No cross reactivity with other acetylated Lysines in histone H3.
Western Blot: 0.5 ug/mL - 2 ug/mL;
ICC: 0.5 ug/mL - 2 ug/mL;
ChIP: 2 ug/mL-10 ug/mL;
IHC: 1 ug/mL-10 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.