Anti-H3K36me3 Rabbit Monoclonal Antibody [clone: RM155]
Catalog # 103255-452
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Primary
- Antigen Name:Trimethylated Histone H3 Lysine 36, H3K36me3
- Antigen Symbol:H3K36me3
- Clonality:Monoclonal
- Clone:Clone: RM155
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:100 µg
- Epitope:Trimethyllysine 36 in Histone H3
- ChIP:Yes
- Format:Purified
- Form:Liquid
- Gene ID:8350
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Concentration:1 mg/mL
- Shipping Temperature:4°C
- Immunogen:A trimethyl-peptide corresponding to trimethyl-Histone H3 (Lys36).
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-452
- Supplier no.:31-1051-00
Specifications
About this item
This antibody reacts to Histone H3 trimethylated at Lysine 36 (K36me3). No cross reactivity with monomethylated Lysine 36 (K36me1) or dimethylated Lysine 36 (K36me2), or other methylations in histone H3.
Western Blot: 0.5 ug/mL – 2 ug/mL;
ChIP: 2 ug/mL- 10 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.