Order Entry
United States
ContactUsLinkComponent
Anti-ACTB Rabbit Monoclonal Antibody [clone: RM112]
Anti-ACTB Rabbit Monoclonal Antibody [clone: RM112]
Catalog # 103255-438
Supplier:  RevMAb Biosciences
undefined
Anti-ACTB Rabbit Monoclonal Antibody [clone: RM112]
Catalog # 103255-438
Supplier:  RevMAb Biosciences
Supplier Number:  31-1013-00

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Actin beta
  • Antigen Symbol:
    ACTB
  • Clonality:
    Monoclonal
  • Clone:
    Clone: RM112
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Multiple
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Epitope:
    N-terminal
  • Format:
    Purified
  • Form:
    Liquid
  • Gene ID:
    60
  • Antigen Synonyms:
    beta-actin
  • Storage Buffer:
    50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
  • Storage Temperature:
    -20.0°C
  • Shipping Temperature:
    4°C
  • Immunogen:
    A peptide corresponding to the N-terminus of β-actin
  • Purification:
    Protein A affinity purified from an animal origin-free culture supernatant
  • Cat. No.:
    103255-438
  • Supplier no.:
    31-1013-00

Specifications

About this item

This antibody reacts to β-Actin (Actin, cytoplasmic 1). No cross reactivity with other isoforms of Actin.

Western Blot (WB): 1:1000 dilution;
Immunoprecipitation (IP): 1:200 dilution;
Chromatin IP (ChIP): 1:200 dilution;
Immunocytochemistry (ICC): 1:200 dilution;
Immunohistochemistry (IHC): 1:200 dilution.