Anti-ACTB Rabbit Monoclonal Antibody [clone: RM112] (Biotin)
Catalog # 103255-224
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Secondary
- Antigen Name:Actin beta
- Antigen Symbol:ACTB
- Clonality:Monoclonal
- Clone:Clone: RM112
- Conjugation:Biotin
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:50 µg
- Epitope:N-terminal
- Format:Purified
- Form:Liquid
- Gene ID:60
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Shipping Temperature:4°C
- Immunogen:A peptide corresponding to the N-terminus of β-actin
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-224
- Supplier no.:31-1013-02
Specifications
About this item
This antibody reacts to β-Actin (Actin, cytoplasmic 1). No cross reactivity with other isoforms of Actin.
Western Blot (WB): 1:1000 dilution;
Immunoprecipitation (IP): 1:200 dilution;
Chromatin IP (ChIP): 1:200 dilution;
Immunocytochemistry (ICC): 1:200 dilution;
Immunohistochemistry (IHC): 1:200 dilution.