Order Entry
United States
ContactUsLinkComponent
Lightning in a Bottle™, Chemiluminescent Membrane/Blotting Peroxidase Substrate
Lightning in a Bottle™, Chemiluminescent Membrane/Blotting Peroxidase Substrate
Supplier:  Immuno4 LLC
Certificates
Lightning in a Bottle™, Chemiluminescent Membrane/Blotting Peroxidase Substrate
Supplier:  Immuno4 LLC

Specifications

  • Form:
    Liquid
  • Shelf Life:
    18 Months at 2° to 8°C
  • Storage Temperature:
    2° to 8°C
  • Application:
    Membrane Immunoassay, Blotting, Western Blotting
  • Color:
    Colorless, Clear

Specifications

Frequently Bought Together