Lumi Super Tracker™ Soluble/ELISA Chemiluminescent Peroxidase Substrate, ImmunO4
Supplier: Immuno4 LLC
Certificates
About this item
Developed specifically for ELISA and In-Vitro Diagnostic applications requiring ultra sensitivity with exceptional signal to noise ratios.
- Ultimate sensitivity with true low-femto detection
- Low backgrounds
- Highest signal to noise ratios
- Superior Stability
- 18-month shelf life
LUMI Super Tracker™ is one of the best chemiluminescent substrates on the market
ImmunO4 stabilized Lumi Super Tracker Luminol based substrates can detect horseradish peroxidase at the femtogram level. ImmunO4 chemiluminescent substrates provide superior sensitivity compared to competitor products. These reagents are stable for several days after mixing. The substrate is supplied as two components (1:1).
Great sensitivity with very low background.
Specifications
- Form:Liquid
- Shelf Life:18 Months at 2° to 8°C
- Storage Temperature:2° to 8°C
- Application:Soluble Immunoassay, ELISA
- Color:Colorless, Clear
Specifications
Frequently Bought Together