Super Shield™ Macro™ Universal Protein Stabilizer, ImmunO4
Supplier: Immuno4 LLC
Certificates
About this item
Preserves and stabilizes the activity of various biomolecules used in immunoassays and molecular assays.
- No Mammalian proteins
- Minimizes background interactions and issues
- Eliminates BSE regulatory concerns
- ImmunO4 MACZR™ is compatible and works well with ImmunO4 Coat Block Diluents
- Can be used as a stabilizer, buffer and diluent
- Room temperature stable reagent
- Stable through freeze cycles
- Superior all-purpose stabilizer
This universal stabilizer was developed to maximize the life of various biomolecules while eliminating non-specific protein interactions
MACZR™ is designed as a ready-to-use buffer and stabilizer for immunoassays and molecular assays. It can be used for high concentration stock and dilute biomolecule solutions. Super ShieldTM can be used directly as an assay buffer. Super Shield™ is stable and can be used at room temperature or 2° to 8°C. It stabilizes dilute biomolecules for months at room temperature.
The stabilizer is supplied as a one component liquid, ready-to-use formula.
Specifications
- Form:Liquid
- Shelf Life:24 Months at 2° to 8°C
- Storage Temperature:2° to 8°C
- Application:Immunoassay, ELISA, Blotting
- Color:Pale Yellow to Tan
Specifications
Frequently Bought Together