Order Entry
United States
ContactUsLinkComponent
Super Shield™ Alkaline Phosphatase (AP) Conjugate and Protein Stabilizer, ImmunO4
Super Shield™ Alkaline Phosphatase (AP) Conjugate and Protein Stabilizer, ImmunO4
Supplier:  Immuno4 LLC
Certificates
Super Shield™ Alkaline Phosphatase (AP) Conjugate and Protein Stabilizer, ImmunO4
Supplier:  Immuno4 LLC

Specifications

  • Form:
    Liquid
  • Shelf Life:
    24 Months at 2° to 8°C
  • Storage Temperature:
    2° to 8°C
  • Application:
    Immunoassay, ELISA, Blotting
  • Color:
    Pale Yellow to Tan

Specifications

Frequently Bought Together