Adjustable Flow Control Valve, Allegro
57006-056
: Allegro
Small Business Enterprise
- Cat. No.:57006-056
- Pressure range:20 - 45 psig
- Supplier no.:9992
The high pressure Adjustable Flow Control Valve adjusts air flow from any high pressure air source for greater comfort.
- Large Gripping Handle
- Adjustable Air Flow from Any High-Pressure Air Source
- Adjustable with Gloved Hands
It offers a large gripping handle that makes it easy to use with a gloved hand.
From the lowest flow position, the control valve will deliver the minimum flow required by NIOSH.