ContactUsLinkComponent
National D-ELS-4 Zoom Stereomicroscope
National D-ELS-4 Zoom Stereomicroscope
Catalog # 470222-460
Supplier:  National Optical
undefined
National D-ELS-4 Zoom Stereomicroscope
Catalog # 470222-460
Supplier:  National Optical
Supplier Number:  D-ELS-4
Specifications
  • Description:
    Stereomicroscope 0.7X - 4.5X Zoom
  • Illumination:
    LED Cordless
  • Magnification:
    7X to 45X
  • Cat. No.:
    470222-460
  • Supplier no.:
    D-ELS-4
Specifications
About this item
The D-ELS-4 model is a new stereo-zoom microscope perfect for high school students.
  • Zoom: 1:6
  • Magnification: 0.7x–4.5x
  • Dual LED Energy-Efficient Illumination Lasts up to 50,000 Hours
  • Three-Year Limited Manufacturer's Warranty
  • Designed for High School Classes
It allows students to view objects at .7x magnification then zoom to 45x magnification for an exciting, close-up view.
The microscope has energy-efficient LED illumination that lasts up to 50,000 hours.
Ordering information: This product includes a manufacturer's warranty.