Boreal Digital Research Microscope with Tablet
Catalog # 470222-456
Supplier: Avantor
Specifications
- Description:Binocular Microscope with 8" LCD Tablet
- Environmentally Preferable:
- Illumination:LED Corded
- Magnification:4X, 10X, 40X, 100X Objectives
- Stage:Rackless Stage
- Cat. No.:470222-456
- Supplier no.:BTW1-169-SP
Specifications
About this item
The Tablet Series incorporates a wide range of features designed for university and laboratory use.
- Pre-Loaded with Motic Apps to View and Share Live Images
- Eight-Inch LCD Android Tablet with 1280 x 800 Screen Resolution
- Mini-HDMI Output for High-Resolution Viewing
- Built-In Motic Hub for Wireless Sharing
Superb optical clarity and reliable mechanical engineering assure years of top performance.
Like the MoticCam X, this Wi-Fi digital tablet transmits live images to iOS or Android devices. Use it as a conventional microscope or share live images with colleagues using Wi-Fi tablets, wireless laptops, and HD-ready monitors/projectors via HDMI. The tablet includes preloaded Motic apps.