Order Entry
United States
Orders LinkContactUsLinkComponent
Human Native LDL (from Plasma)

Human Native LDL (from Plasma)

100 µg
Catalog # 102964-842
Marketsource
Supplier:   Cell Biolabs
Supplier #:  STA-214
Human Native LDL (from Plasma)
Catalog # 102964-842
Supplier:  Cell Biolabs
Marketsource
CAS Number:  
Human Native LDL (from Plasma)
Human Native LDL (from Plasma)

100 µg

CAS Number:  
Price
$473.85
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Documents

Specifications

Specifications

  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Cat. No.:
    102964-842
  • Protein/Peptide Type:
    Native
  • Purity:
    98% by SDS-PAGE
  • Protein/Peptide Name:
    LDL (Copper (Cu++) oxidized)
  • Species:
    Human
  • Source:
    Plasma

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.