Order Entry
United States
ContactUsLinkComponent
Difco™ Triple Sugar Iron Agar, BD Biosciences
Difco™ Triple Sugar Iron Agar, BD Biosciences
Catalog # 90001-896
Supplier:  BD
CAS Number:  
Difco™ Triple Sugar Iron Agar, BD Biosciences
Catalog # 90001-896
Supplier:  BD
Supplier Number:  226540
CAS Number:  

Specifications

  • Description:
    Difco™ Triple Sugar Iron Agar
  • Size:
    500 g
  • Cat. No.:
    90001-896
  • Supplier no.:
    226540

Specifications

About this item

Triple Sugar Iron Agar (TSI Agar) is used for the differentiation of Gram-negative enteric bacilli based on carbohydrate fermentation and the production of hydrogen sulfide.

  • Contains three sugar: dextrose, lactose, and sucrose
  • Phenol red for detecting carbohydrate fermentation
  • Ferrous ammonium sulfate detects hydrogen sulfide production
  • Dehydrated appearance: Pink, free-flowing

The addition of sucrose to the double sugar formulation of Kligler Iron Agar increased the sensitivity of the medium by facilitating the detection of sucrose-fermenting bacilli, as well as lactose and/or dextrose fermenters.