About this item
Todd Hewitt Broth is a general-purpose medium, which primarily is used for the cultivation of beta-hemolytic strepto-cocci, especially for serological studies.
- Contains peptones, dextrose, and salts for promoting general growth
- Dehydrated apperarance: Light beige, free-flowing
Selective enrichment broths include the advantages of both enrichment and selection by providing conditions conducive to the growth of group B streptococci while inhibiting the growth of contaminants.
2 Options Available Below
Product Details & Documents
Todd Hewitt Broth is a general-purpose medium, which primarily is used for the cultivation of beta-hemolytic strepto-cocci, especially for serological studies.
- Contains peptones, dextrose, and salts for promoting general growth
- Dehydrated apperarance: Light beige, free-flowing
Selective enrichment broths include the advantages of both enrichment and selection by providing conditions conducive to the growth of group B streptococci while inhibiting the growth of contaminants.
Documents
Product Family Options
Product Information