ContactUsLinkComponent
Difco™ Thioglycollate Medium without Indicator, BD Life Sciences
Difco™ Thioglycollate Medium without Indicator, BD Life Sciences
Catalog # 90003-290
Supplier:  BD
CAS Number:  
Difco™ Thioglycollate Medium without Indicator, BD Life Sciences
Catalog # 90003-290
Supplier:  BD
Supplier Number:  243010
CAS Number:  
Specifications
  • Description:
    Difco™ Thioglycollate Medium without Indicator
  • Size:
    500 g
  • Cat. No.:
    90003-290
  • Supplier no.:
    243010
Specifications
About this item
Thioglycollate Medium without Indicator is a general-purpose medium for the cultivation of microorganisms, especially obligate anaerobes.
  • Casein and soybean peptones, dextrose, and cystine supply nitrogen and carbon
  • Sodium chloride provides essential ions
Casein and soybean peptones, dextrose, and cystine supply nitrogenous and carbonaceous compounds, fermentable carbohydrates, and trace ingredients. This medium is satisfactorily used as an enrichment culture for various types of specimens and also as a transportation medium.