Difco™ Supplement C, BD Biosciences
Catalog # 90003-468
Supplier: BD
CAS Number:
Specifications
- Description:Difco™ Supplement C
- Size:5 mL
- Cat. No.:90003-468
- Supplier no.:252710
Specifications
About this item
Supplement C is a sterile yeast concentrate for use in the isolation of fastidious microorganisms.
- Yeast concentrate
- Sterile
- Suitable for use with fastidious microorganisms
Supplement C is a yeast concentrate for use in supplementing media for growth of microorganisms with exacting nutritional requirements.