ContactUsLinkComponent
Difco™ Sodium Desoxycholate, BD Life Sciences
Difco™ Sodium Desoxycholate, BD Life Sciences
Catalog # 90001-806
Supplier:  BD
CAS Number:  
Difco™ Sodium Desoxycholate, BD Life Sciences
Catalog # 90001-806
Supplier:  BD
Supplier Number:  224820
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Difco™ Sodium Desoxycholate
  • Size:
    100 g
  • Environmentally Preferable:
  • Cat. No.:
    90001-806
  • Supplier no.:
    224820
Specifications
About this item
Sodium salt of desoxycholic acid used for the pneumococcus bile solubility test and as a media component.
  • Popular anionic, bile-acid detergent
  • Can be removed from solution by dialysis
  • High-purity compound with low UV absorptivity
Sodium desoxycholate is an ionic detergent that is especially useful for disrupting and dissociating protein interactions.