Order Entry
United States
ContactUsLinkComponent
Rappaport-Vassiliadis (MSRV) Semisolid Modification, BD Diagnostics
Rappaport-Vassiliadis (MSRV) Semisolid Modification, BD Diagnostics
Catalog # 90001-020
Supplier:  BD
CAS Number:  
Rappaport-Vassiliadis (MSRV) Semisolid Modification, BD Diagnostics
Catalog # 90001-020
Supplier:  BD
Supplier Number:  218681
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Rappaport-Vassiliadis (MSRV) Medium, Semisolid Modification
  • Size:
    500 g
  • Environmentally Preferable:
  • Cat. No.:
    90001-020
  • Supplier no.:
    218681

Specifications

About this item

Rappaport-Vassiliadis Medium, Semisolid Modification is used with Novobiocin Antimicrobic Supplement in rapidly detecting motile Salmonella in feces and food products

  • Contains peptones as carbon and nitrogen sources for general growth requirements
  • Novobiocin supplement helps inhibit organisms other than Salmonella
  • Low pH of the medium helps to select for highly resistant Salmonella species
  • Agar is the solidifying agent

Use with Novobiocin Antimicrobic Supplement.