ContactUsLinkComponent
Extension Set, Baxter IV Solutions
Extension Set, Baxter IV Solutions
Catalog # 68000-878
Supplier:  Baxter
undefined
Extension Set, Baxter IV Solutions
Catalog # 68000-878
Supplier:  Baxter
Supplier Number:  2C5641
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Volume:
    0.87 ml
  • Description:
    Extension Set with Male Luer Lock Adapter and Retractable Collar
  • Cat. No.:
    68000-878
  • Supplier no.:
    2C5641
Specifications
About this item
Extension set with Male Luer Lock connector.
  • Contains DEHP
  • Single use only
  • Fluid path is sterile, nonpyrogenic