Cynomolgus Monkey Serum, Equitech- Bio, Inc.
Supplier: Avantor
Certificates
About this item
Collected from sexually-mature donor cynomolgus monkeys that are free of common parasites.
- Sterility tested and filtered
- Lot-to-lot consistency
- Heat inactivation available
Subject to CITES regulations. Sterile-Filtered serum is sterility tested and sterile-filtered to 0.2 micron absolute. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Heat inactivation is available if desired.
Frequently Bought Together