About this item
These Label IT® siRNA Tracker Intracellular Localization Kits are efficient, direct labeling for in vitro and in vivo tracking of siRNA.
- Labels 50 µg siRNA
- Easily and consistently control the labeling reactions and labeling density
- High efficiency labeling with optimal visualization of siRNA in cells and tissues
Generate a labeled siRNA sample without hindering its inherent ability to silence targeted genes. Monitor both sub-cellular localization and functional inhibition of target gene expression simultaneously following introduction of the labeled siRNA into mammalian cells. Contain sufficient reagents to label 50 µg of siRNA. siRNA can be delivered to cells in vitro or in vivo using the transfection reagent of your choice.
Versatile Labeling – Efficiently label and visualize the siRNA of your choice.
One-step chemical method – Easily and consistently control the labeling reactions and labeling density.
High efficiency labeling – Optimal visualization of siRNA in cells and tissues.