Label IT® Plasmid Delivery Controls, Mirus Bio
Supplier: Mirus Bio
Certificates
About this item
These fluorescently labeled plasmids are ideal for in vitro and in vivo tracking.
- Easily visualize transfected cells and assess delivery efficiency using fluorescent microscopy
- Suitable for co-delivery experiments with functional plasmids
Sensitive - Easily visualize transfected cells and assess delivery efficiency using fluorescent microscopy.
Compatible - Suitable for co-delivery experiments with functional plasmids.
Ease of use - Supplied as a ready to use 0.5 mg/ml concentration for both in vitro and in vivo tracking studies.
Delivery: Supplied as a ready to use 0.5 mg/ml concentration for both in vitro and in vivo tracking studies.