- Description:Malchite Green Phosphate Assay Kit
- Environmentally Preferable:
- Storage Temperature:4°C
- Cat. No.:10634-098
- Supplier no.:KA1331
Malachite green assay kit provides a fast, reproducible, and non-radioactive method for measuring inorganic free phosphate in aqueous solutions
These are based on the complex formed between malachite green molybdate and free orthophosphate under acidic conditions.