BBL™ BCYE Agar Base, BD
Catalog # 90000-604
Supplier: BD
CAS Number:
Specifications
- Description:BBL™ BCYE Agar Base
- Size:500 g
- Cat. No.:90000-604
- Supplier no.:212327
Specifications
About this item
BCYE Agar Base is used for the selective cultivation of Legionella species.
- Highly selective medium
- Supplemented with cephalothin, colistin, vancomycin, and cycloheximide
Activated charcoal decomposes hydrogen peroxide and other toxic products; ACES Buffer was added to formulation to get a stable pH for optimal growth.