Order Entry
United States
Orders LinkContactUsLinkComponent
Luria Agar Base, BD

Luria Agar Base, BD

Luria Agar Base, 500 g
Catalog # 90003-250
Supplier:   BD
Supplier #:  241320
Luria Agar Base, BD
Catalog # 90003-250
Supplier:  BD
CAS Number:  
Luria Agar Base, BD

Luria Agar Base, 500 g

CAS Number:  
Price
$216.81
New Customer Promotional Price
Log in for availability
Each (500g)
Product Details & Documents

Luria Agar is used for routine cultivation and estimation of not particularly fastidious microorganisms.

  • Firm, comparable with 1.5% Agar gel
  • Cream to yellow homogenous free-flowing powder
  • Nutritionally rich

Luria Agar is prepared as described by Lennox for cultivation and maintenance of recombinant strains of Escherichia coli. This media is generally used for molecular and genetic studies because of its nutritive capacity and simple composition, which can be easily altered as per specific requirements.

Documents

Specifications

Specifications

  • Description:
    Luria Agar Base
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.