Tegaderm™ Transparent Film Roll, 3M™
Supplier: Healthcare Products
Certificates
About this item
Nonsterile roll version of Tegaderm™ Transparent Film Dressing provides all the benefits of surgical tape while providing a waterproof, bacterial, and viral barrier.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.