Solus™ Protective Eyewear with Anti-Fog Lens, 1000 Series, 3M™
Supplier: 3M
Certificates
About this item
Solus™ 1000 Series protective eyewear features Scotchgard™ Anti-Fog lens coating, which helps workers see more clearly over a longer duration.
- Removable temples enable the use of the optional strap
- Optional foam gasket heps reduce eye exposure
- Available in several frame colors and lens tints
- All lenses offer 99.9% UV protection
The coating provids longer lasting fog resistance than traditional anti-fog coatings.