SecureFit™ Protective Eyewear, SF400 Series, 3M™
Supplier: 3M
Certificates
About this item
SecureFit™ SF400 Series eyewear features soft adjustable nose pads, dual injected temples, and 3M™ Pressure Diffusion Temple Technology to help diffuse pressure over the ear.
- Eyewear design enhances frame comfort without compromising the security of fit across a diverse workforce
- All lenses offer 99.9% UV protection