Solus™ 1000 Series Protective Eyewear Kit, 3M™
Supplier: 3M
Certificates
About this item
Solus™ 1000 Series Protective Eyewear Kit features Scotchgard™ Anti-Fog lens coating, which helps workers see clearly for longer durations.
- Removable temples enable the use of the optional strap
- Optional foam gasket heps reduce eye exposure
- All lenses offer 99.9% UV protection
- Available in multiple frame colors and lens tints
The coating provides longer lasting fog resistance than traditional anti-fog coatings.
Frequently Bought Together