gWIZ Mammalian Expression Vectors, Genlantis
Supplier: Avantor
Certificates
About this item
The gWiz™ high expression vector has been systematically engineered to produce the highest levels of reporter gene expression in a wide variety of mammalian cells and tissues.
- High plasmid yield in E.coli
- High levels of transient gene expression
- Suitable for in vitro and in vivo gene expression studies applications
The gWiz vector obtains its exceptional gene expression levels from an optimized human cytomegalovirus (CMV) promoter followed by intron A from the CMV immediate-early (IE) gene. In addition, the gWiz vector contains an optimized plasmid backbone that has been extensively altered to achieve abundant plasmid production in E.coli.
Frequently Bought Together