Order Entry
United States
ContactUsLinkComponent
DNA (Deoxyribonucleic acid) 10 mg/ml in water (from fish sperm), sonicated
DNA (Deoxyribonucleic acid) 10 mg/ml in water (from fish sperm), sonicated
Catalog # 10639-332
Supplier:  Abnova
CAS Number:  
DNA (Deoxyribonucleic acid) 10 mg/ml in water (from fish sperm), sonicated
Catalog # 10639-332
Supplier:  Abnova
Supplier Number:  R0014
CAS Number:  

Specifications

  • Size:
    5×1 ml
  • Cat. No.:
    10639-332
  • Supplier no.:
    R0014

Specifications

About this item

 :
Storage Temperature:Freezer
CAS Number:9007-49-2
 :73049-39-5
MDL Number:MFCD00130922