Versaflo™ Healthcare Powered Air Purifying Respirator (PAPR) Assembly, 3M™
10837-172
: 3M
- Description:Versaflo™ Healthcare PAPR Assembly (magenta), belt-mounted
- Battery Type:Standard Capacity Lithium Ion, Rechargeable
- Cat. No.:10837-172
- Supplier no.:51131275027
Workers can customize their respiratory protection system by choosing the headtop.
- Intended for use in Pharmaceutical and Healthcare industries
With the option to customize, workers can design a respiratory system that meets their specific needs.
: NIOSH Approved.
: System includes PAPR Unit, Standard Battery, Single-Station Battery Charger, Length Adjusting Breathing Tube, (HE) Particulate Filter, Filter Cover, Easy Clean Belt, Airflow Indicator, Cleaning and Storage Kit, and Storage Bag.