Order Entry
United States
ContactUsLinkComponent
Steri-Drape™ Urology Drape, 3M™
Steri-Drape™ Urology Drape, 3M™
Catalog # 75787-234
Steri-Drape™ Urology Drape, 3M™
Catalog # 75787-234
Supplier Number:  MMM 1071

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Urology Drape, with 2" Circle Aperture & Finger Cot with Adhesive
  • Size:
    58.4 x 241 cm (23 x 95")
  • Cat. No.:
    75787-234
  • Supplier no.:
    MMM1071

Specifications

About this item

Plastic specialty drape features an aperture and a built-in finger cot for use in urology and gynecology surgical procedures.