EnviroTrans™ Swab Rinse Kit, Barney Miller Broth, Hardy Diagnostics
Catalog # 75813-998
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Barney Miller Broth
- Cat. No.:75813-998
- Supplier no.:SRK100
Specifications
About this item
EnviroTrans™ Swab Rinse Kit, Barney Miller Broth is recommended for environmental monitoring.
- Collect and transport environmental samples with efficiency and ease.
- Prefilled with Barney Miller Broth
Recommended for environmental monitoring.
Collect and transport environmental samples with efficiency and ease. Simply remove the cap and attached Dacron® swab, collect the sample, recap, and send to the lab. Each EnviroTrans™ tube contains broth or buffer.