Quidel QuickVue® Dipstick Strep A test, controls included, Hardy Diagnostics
Catalog # 75813-942
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:QuickVue, Dipstick Strep A
- Size:50 tests
- Cat. No.:75813-942
- Supplier no.:20108
Specifications
About this item
QuickVue® Dipstick Strep A test, intended for use as an aid in the diagnosis of Group A Streptococcal infection.
- Results in five minutes or less
- 95% overall agreement with SBA culture
- High sensitivity at low levels of colony count
- CLIA waived
Dipstick immunoassay for the rapid qualitative detection of Group A Streptococcal antigen from throat swab specimens.