Order Entry
United States
ContactUsLinkComponent
Ruthenium ICP Standard, Inorganic Ventures
Ruthenium ICP Standard, Inorganic Ventures
Catalog # 100360-642
Supplier:  Inorganic Ventures
CAS Number:  
undefined
Ruthenium ICP Standard, Inorganic Ventures
Catalog # 100360-642
Supplier:  Inorganic Ventures
Supplier Number:  CGRU1-500ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    10% HCl
  • Color:
    Clear/Amber
  • Size:
    500 ml
  • Cat. No.:
    100360-642
  • Element:
    Ruthenium
  • Supplier no.:
    CGRU15

Specifications

About this item

Certified reference material for Ruthenium, traceable to NIST.

One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).