ContactUsLinkComponent
Glucagon-Like Peptide-1, GLP-1, Human ELISA, Multispecies specificity, BioVendor
Glucagon-Like Peptide-1, GLP-1, Human ELISA, Multispecies specificity, BioVendor
Catalog # 101424-328
Supplier:  BioVendor
CAS Number:  
undefined
Glucagon-Like Peptide-1, GLP-1, Human ELISA, Multispecies specificity, BioVendor
Catalog # 101424-328
Supplier:  BioVendor
Supplier Number:  YK160
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Primary antibody reactivity:
    Human
  • Description:
    Glucagon-Like Peptide-1 (GLP-1) Human ELISA (Multispecies specificity)
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    101424-328
  • Supplier no.:
    RSCYK160R
Specifications
About this item
GLP-1 is a peptide hormone from the intestinal mucosa, which is produced from its precursor, proglucagon by post transnational processing.
  • Calibration Range: 0.206–50 ng/mL
  • Plasma: 30 uL/well
GLP-1 is a peptide hormone from the intestinal mucosa, which is produced from its precursor, proglucagon by post transnational processing. The mammalian proglucagon 1) is synthesized in the neuroendocrine L-cell of the intestine and the alpha-cells of the pancreas. It contains within its structure the sequences of glucagon and two glucagon-like peptides (GLP-1 and GLP-2) in tandem flanked at their amino and carboxyl termini by dibasic residues. GLP-1 is a 37 amino acids peptide and produced in the small intestine and in the pancreas in the human, in either C-terminal-amidated on glycine-extended form. GLP1 (7–36) amide and its receptor are present in several brain regions and may play a role in the physiological control of feeding4). Several reports have been presented as follows as to the biological activities of GLP-1. GLP-1 (7–37) and (7–36) amide is known as one of the most potent insulin secretagogues. GLP-1 (7–36) amide was supposed to improved glycemic control in patients with type 2 diabetes by increasing insulin secretion, by inhibiting glucagon secretion and by delaying gastric emptying rather than by altering extrapancreatic glucose metabolism. Intravenous GLP-1(7–37) and (7–36)amide could normalize fasting hyperglycaemia in type 2 diabetic patients. Hyperglycaemia during parenteral nutrition could be controlled by exogenous GLP-1, whereas the chronic therapy of type 2 diabetes required GLP-1 derivatives with longer duration of action. Recombinant GLP-1 (7–36) amide was recently shown to cause significant weight loss in type 2 diabetics when administered for 6 weeks as a continuous subcutaneous infusion, 5-day treatment of hereby obese human subjects with GLP-1 at high doses by prandial subcutaneous infusion promptly slowed gastric emptying as a probable mechanism of action of increased satiety, decreased hunger and reduced food intake with an ensuing weight loss. A G-protein-coupled receptor, GPR120, which is abundantly expressed in intestine, functions as a receptor for unsaturated long-chain FFAs (free fatty acids). The stimulation of GPR120 by FFAs promotes the secretion of GLP-1 in vitro (measured by YK160, Yanaihara Institute Inc) and in vivo, and increases circulation insulin, indicate that GPR120-mediated GLP-1 secretion induced by dietary FFAs is important in the treatment of diabetes.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.