Specifications
- Target protein:TNF-alpha
- Description:TNF-alpha Total Monkey ELISA
- Size:1 kit
- Regulatory Status:RUO
- Cat. No.:102552-612
- Supplier no.:RAF131R
Specifications
About this item
TNF-α (17.5 kDa, 157 amino acid) is a non-glycosylated protein that is a potent lymphoid factor, which exerts cytotoxic effects on a wide range of tumor cells and other target cells.
- Calibration Range: 28.1-1800 U/mL
- Limit of Detection: 6 U/mL
- Baboon serum, Cell culture supernatant, Cynomolgus serum: 50 µL/well
TNF-α (17.5 kDa, 157 amino acid) is a non-glycosylated protein that is a potent lymphoid factor, which exerts cytotoxic effects on a wide range of tumor cells and other target cells. It is secreted by macrophages, monocytes, neutrophils, T-cells and NK-cells following their stimulation by bacterial lipopolysaccharides. TNF-α has been suggested to play a pro-inflammatory role and has been detected in synovial fluid of patients with rheumatoid arthritis.