Order Entry
United States
ContactUsLinkComponent
sTNF-R I Monkey ELISA, BioVendor
sTNF-R I Monkey ELISA, BioVendor
Catalog # 102552-622
Supplier:  BioVendor
CAS Number:  
undefined
sTNF-R I Monkey ELISA, BioVendor
Catalog # 102552-622
Supplier:  BioVendor
Supplier Number:  RAF116R
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Monkey
  • Target protein:
    sTNF-R
  • Description:
    sTNF-R I Monkey ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-622
  • Supplier no.:
    RAF116R

Specifications

About this item

This assay will help to clarify the possible diagnostic and prognostic value of circulating sTNF-R (60 kDa) in various neoplastic and inflammatory diseases.

  • Calibration Range: 0.09-6 U/mL
  • Limit of Detection: 0.5 U/mL
  • Baboon serum: 10 µL/well

Tumor Necrosis Factor (TNF) was originally discovered in sera of animals and was found to cause hemorrhagic necrosis of some transplantable mouse and human tumors and to exhibit primarily cytotoxic activities against tumor but not normal cells in vitro. The TNF family consists of two proteins designated TNFα, also called cachectin, and TNFβ, also called lymphotoxin, which are pleiotropic cytokines that can mediate a wide variety of biological effects.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.